missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RSRC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16330855
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16330855 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16330855 Supplier Invitrogen™ Supplier No. PA583899

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ARRLERAKKL QEQREKEMVE KQKQQEIAAA AATGGSVLNV AALLASGTQV TPQIAMAAQM AALQAKALAE TGIAVPSYYN PAAVNPMKFA EQEKKRKMLW QG Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.

In vitro study revealed that RSRC2 might play a role in cell proliferation. RSRC2 may be a novel tumor suppressor of esophageal cancer cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSRC2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RSRC2
Gene Accession No. Q7L4I2
Gene Alias 1500011J06Rik; arginine and serine rich coiled-coil 2; arginine/serine-rich coiled-coil 2; arginine/serine-rich coiled-coil protein 2; arginine/serine-rich coiled-coil protein 2-like protein; FLJ11021; Rsrc2
Gene Symbols RSRC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RSRC2. Recombinant protein control fragment (Product #RP-104850).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65117
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.