missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSU1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94442-0.02ml
This item is not returnable.
View return policy
Description
RSU1 Polyclonal antibody specifically detects RSU1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RSU1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ31034, Ras suppressor protein 1, ras suppressor protein 1 variant 1, ras suppressor protein 1 variant 2, RSP1, RSP-1ras suppressor protein 1 variant 3, Rsu-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-277 of human RSU1 (NP_036557.1). MSKSLKKLVEESREKNQPEVDMSDRGISNMLDVNGLFTLSHITQLVLSHNKLTMVPPNIAELKNLEVLNFFNNQIEELPTQISSLQKLKHLNLGMNRLNTLPRGFGSLPALEVLDLTYNNLSENSLPGNFFYLTTLRALYLSDNDFEILPPDIGKLTKLQILSLRDNDLISLPKEIGELTQLKELHIQGNRLTVLPPELGNLDLTGQKQVFKAENNPWVTPIADQFQLGVSHVFEYIRSETYKYLYGRHMQANPEPPKKNNDKSKKISRKPLAAKNR | |
| 0.02 mL | |
| Vision | |
| 6251 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction