missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSV/BRSV Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93585AF532
This item is not returnable.
View return policy
Description
RSV/BRSV Polyclonal antibody specifically detects RSV/BRSV in Virus samples. It is validated for Western Blot, Flow Cytometry
Specifications
| RSV/BRSV | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, Flow Cytometry | |
| Alexa Fluor 532 | |
| Bovine Respiratory Syncytial Virus BRSV, Bovine RSV | |
| Residues 201-211 [KELLPKVNNHDC] of the 63 kDa BRSV F protein. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction