missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSV M Protein Recombinant Rabbit Monoclonal Antibody (HL3000)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556856
This item is not returnable.
View return policy
Description
RSV M Protein Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The M protein of the Respiratory Syncytial Virus (RSV) is a crucial structural protein involved in virus assembly and budding. This matrix protein plays a central role in coordinating the assembly of viral components at the plasma membrane, facilitating the formation of infectious virions. The M protein interacts with the viral envelope glycoproteins and the nucleocapsid, ensuring the proper encapsidation of the viral RNA and the release of mature virions from the host cell. Due to its essential function in the viral life cycle, the RSV M protein is a significant target for antiviral drug development. Inhibiting the M protein can potentially disrupt the assembly and release of RSV, thereby reducing viral replication and spread. Research on the RSV M protein aims to understand its interactions and mechanisms further, which could lead to the development of effective therapeutic interventions against RSV infections.
Specifications
| RSV M Protein | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL3000 | |
| Unconjugated | |
| human respiratory syncytial virus; RSV | |
| Full length Respiratory Syncytial Virus M protein. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction