missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RSV Monoclonal Antibody (RSV 3132)
GREENER_CHOICE

Product Code. 13451017
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13451017 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13451017 Supplier Invitrogen™ Supplier No. MA516283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RSV Monoclonal Antibody for ICC/IF

Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV, Long, Randall, RSV Strains 127, SNK & 9007
Applications Immunocytochemistry
Classification Monoclonal
Clone RSV 3132
Concentration 6.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 15 mM sodium azide; pH 7.5
Gene Alias human respiratory syncytial virus; RSV
Host Species Mouse
Immunogen RSV strains 127, SNK & 9007, Long, Randall, 8/60 & A/2.
Purification Method Protein A
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.