missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSV NP Recombinant Rabbit Monoclonal Antibody (HL1296)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550310
This item is not returnable.
View return policy
Description
This product can be used in Sandwich ELISA to detect RSV NP using MA5-46972 as a capture antibody and MA5-50310 as the detection antibody.
Specifications
| RSV NP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| HL1296 | |
| Unconjugated | |
| human respiratory syncytial virus; RSV; RSV NP; RSV Nuclear Protein | |
| Recombinant protein encompassing a sequence within the center region of Respiratory Syncytial virus Nucleoprotein. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction