Learn More
RETSCH RT 75 Model Sample Splitter
Product Code. 10421971 Shop All Retsch Products
Click to view available options
Depth (Metric):
75 mm
Unit Size:
Each
Product Code. 10421971
Brand: RETSCH 40.610.0006
Only null left
Specifications
Product Type Sample Splitter
Depth (Metric) 75 mm
Includes 3 Receptacles 8L each, Stand and Dividing Head