missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RT1A [a,b] Rat anti-Rat, BV650, Clone: C3, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 745343
This item is not returnable.
View return policy
Description
RT1A [a,b] Monoclonal antibody specifically detects RT1A [a,b] in Rat samples. It is validated for Flow Cytometry.
Specifications
| RT1A [a,b] | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| Rat | |
| Purified |
| Flow Cytometry | |
| C3 | |
| Brilliant Violet 650 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction