missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTCA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599711
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RTCD1.
RTCD1 catalyzes the conversion of 3'-phosphate to a 2', 3'-cyclic phosphodiester at the end of RNA. The mechanism of action of the enzyme occurs in 3 steps: (A) adenylation of the enzyme by ATP; (B) the enzyme acts on RNA-N3'P to produce RNA-N3'PP5'A; (C) a non catalytic nucleophilic attack by the adjacent 2'hydroxyl on the phosphorus in the diester linkage to produce the cyclic end product. The biological role of this enzyme is unknown but it is likely to function in some aspects of cellular RNA processing. There are two named isoforms.
Specifications
| RTCA | |
| Polyclonal | |
| Unconjugated | |
| RTCA | |
| 2310009A18Rik; AI450277; RNA 3'-terminal phosphate cyclase; RNA cyclase; RNA terminal phosphate cyclase domain 1; RNA terminal phosphate cyclase domain-containing protein 1; RNA-3'-phosphate cyclase; RPC; Rpc1; RTC domain-containing protein 1; RTC1; RTCA; Rtcd1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 66368, 8634 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00442, Q9D7H3 | |
| RTCA | |
| A synthesized peptide derived from human RTCA(Accession O00442), corresponding to amino acid residues V317-L366. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction