Learn More
Invitrogen™ RTCB Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA544610
Product Code. 15586572
  • € 520.00 / 100µL
Description
Peptide sequence: PEAVVSPGGV GFDINCGVRL LRTNLDESDV QPVKEQLAQA MFDHIPVGVG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RTCB
Polyclonal
Unconjugated
Rtcb
3'-phosphate/5'-hydroxy nucleic acid ligase; AI255213; AI463255; ankyrin repeat domain 54; C22orf28; C5H22orf28; D10Wsu52e; DJ149A16.6; FAAP; focal adhesion-associated protein; HAMAP-Rule:MF_03144}; HSPC117; hypothetical protein HSPC117; hypothetical protein LOC406376; hypothetical protein LOC525106; P55; RNA 2',3'-cyclic phosphate and 5'-OH ligase; RNA-splicing ligase RtcB homolog; RTCB; tRNA-splicing ligase RtcB homolog; tRNA-splicing ligase RtcB homolog {ECO:0000255; zgc:76871
Rabbit
Affinity chromatography
RUO
51493
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9Y3I0
Rtcb
Synthetic peptide directed towards the N-terminal of human C22orf28 (aa 107-156).