missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTCB Polyclonal Antibody, Biotin
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142754
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
The function of this protein remains unknown.
Specifications
| RTCB | |
| Polyclonal | |
| Biotin | |
| Rtcb | |
| 3'-phosphate/5'-hydroxy nucleic acid ligase; AI255213; AI463255; ankyrin repeat domain 54; C22orf28; C5H22orf28; D10Wsu52e; DJ149A16.6; FAAP; focal adhesion-associated protein; HAMAP-Rule:MF_03144}; HSPC117; hypothetical protein HSPC117; hypothetical protein LOC406376; hypothetical protein LOC525106; P55; RNA 2',3'-cyclic phosphate and 5'-OH ligase; RNA-splicing ligase RtcB homolog; RTCB; tRNA-splicing ligase RtcB homolog; tRNA-splicing ligase RtcB homolog {ECO:0000255; zgc:76871 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 51493 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9Y3I0 | |
| Rtcb | |
| Peptide with sequence C-QADPNKVSARAKKR. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction