missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTDR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575747
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
RTDR1 (Rhabdoid tumor deletion region protein 1) is a 348 amino acid protein that is expressed in adult spinal cord, skeletal muscle, cerebellum and spleen, as well in some rhabdoid tumors. The gene encoding RTDR1 maps to human chromosome 22, which contains over 500 genes and about 49 million bases.
Specifications
| RTDR1 | |
| Polyclonal | |
| Unconjugated | |
| RSPH14 | |
| 4933431K05Rik; MGC16968; Radial spoke head 14 homolog; radial spoke head homolog 14 (Chlamydomonas); rhabdoid tumor deletion region gene 1; rhabdoid tumor deletion region protein 1; Rsph14; Rtdr1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27156, 365552, 71236 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9D3W1, Q9UHP6 | |
| RSPH14 | |
| Synthetic peptide corresponding to residues in Human RTDR1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction