Learn More
RTEL1 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-22360IR
Product Code. 18136571
  • ONLINE EXCLUSIVE PRICE
    € 625.00
    € 591.15 / 0.10mL
    Save € 33.85
    5% Off
Description
RTEL1 Polyclonal specifically detects RTEL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RTEL1
Polyclonal
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Q9NZ71
RTEL1
A genomic peptide made to an internal region of the human RTEL1 protein (within residues 800-1000). [Swiss-Prot Q9NZ71]
0.1 mL
Cancer, DNA Repair
51750
Store at 4C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
C20orf41, regulator of telomere elongation helicase 1, regulator of telomere length
Rabbit
Affinity Purified
RUO
Primary
Human, Mouse