Learn More
Factor XIIIb Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99806PE
Product Code. 18680759
  • € 407.00 / 0.10mL
Description
Factor XIIIb Polyclonal antibody specifically detects Factor XIIIb in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA
Produced in rabbits immunized with purified, recombinant Human Factor XIIIb (Accession#: P05160; Met1-Thr661)
Antigen and protein A Affinity-purified