Learn More
RTKN Mouse anti-Human, Clone: OTI1A4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505051
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a scaffold protein that interacts with GTP-bound Rho proteins. Binding of this protein inhibits the GTPase activity of Rho proteins. This protein may interfere with the conversion of active, GTP-bound Rho to the inactive GDP-bound form by RhoGAP. Rho proteins regulate many important cellular processes, including cytokinesis, transcription, smooth muscle contraction, cell growth and transformation. Dysregulation of the Rho signal transduction pathway has been implicated in many forms of cancer. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| RTKN | |
| Monoclonal | |
| Unconjugated | |
| RTKN | |
| rhotekin | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6242 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1A4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9BST9 | |
| RTKN | |
| Full length human recombit protein of human RTKN produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.