missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTN1-A/NSP Antibody (MON161), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97678PEATT594
This item is not returnable.
View return policy
Description
RTN1-A/NSP Monoclonal specifically detects RTN1-A/NSP in Human, Mouse, Rat, Hamster samples. It is validated for Flow Cytometry.
Specifications
| RTN1-A/NSP | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| Neuroscience | |
| 6252.0 | |
| Human, Mouse, Rat, Hamster | |
| IgG1 |
| Flow Cytometry | |
| MON161 | |
| Flow Cytometry | |
| RTN1 | |
| MON-161 is a Mouse monoclonal IgG1 antibody derived by fusion of Mouse myeloma cells with spleen cells from a Mouse immunized with a partially purified bacterially expressed Reticulon-1A (NSP-A) hybrid protein (betaGAL-NSP-A 6-776). The epitope of MON-161 | |
| 0.1 mL | |
| Primary | |
| MON-161 exclusively recognizes the 135 kD Reticulon-1A protein in immunoblots of NCI-H82 and other SCLC cell lines, and stains normal and pathological neural and neuroendocrine tissues. | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction