missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12055-2-AP-150UL
This item is not returnable.
View return policy
Description
RTN3 is a member of reticulons, which are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. Multiple promoters rather than alternative splicing of internal exons seem to be involved in this diversity.Specifications
| RTN3 | |
| Polyclonal | |
| Unconjugated | |
| RTN3 | |
| ASYIP, HAP, NSP like protein 2, NSP like protein II, NSPL2, NSPLII, reticulon 3, RTN3, RTN3 A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10313, 140945, 20168 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95197, Q6RJR6, Q9ES97 | |
| RTN3 | |
| RTN3 Fusion Protein Ag2685 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction