missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTN4RL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560710
This item is not returnable.
View return policy
Description
Immunogen sequence: AVPCVSPGLR HGQDLKLLRA EDFRNCTGPA SPHQIKSHTL TTTDRAARKE HHSPHGPTRS KGHPHGPRPG HRKPGKNCTN PRNRNQISKA GAGKQAPELP DYAPDYQHKF SFD Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 76%.
RTN4RL1 may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.
Specifications
| RTN4RL1 | |
| Polyclonal | |
| Unconjugated | |
| Rtn4rl1 | |
| NgR3; Ngrh2; NGRL2; Nogo receptor-like 2; Nogo-66 receptor homolog 2; Nogo-66 receptor-related protein 3; reticulon 4 receptor like 1; reticulon 4 receptor-like 1; reticulon-4 receptor-like 1; RGD:727812}; Rtn4rl1; rtn4rl1 {ECO:0000312 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 146760 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86UN2 | |
| Rtn4rl1 | |
| Recombinant protein corresponding to Human RTN4RL1. Recombinant protein control fragment (Product #RP-88806). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction