missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RTN4RL1 Recombinant Rabbit Monoclonal Antibody (24GB1370)

Product Code. 30275170
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30275170 20 μL 20µL
30276121 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30275170 Supplier Invitrogen™ Supplier No. MA557220

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RTN4RL1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

RTN4RL1 may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTN4RL1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 24GB1370
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rtn4rl1
Gene Accession No. Q80WD0, Q86UN2, Q8K0S5
Gene Alias NgR3; Ngrh2; NGRL2; Nogo receptor-like 2; Nogo-66 receptor homolog 2; Nogo-66 receptor-related protein 3; reticulon 4 receptor like 1; reticulon 4 receptor-like 1; reticulon-4 receptor-like 1; RGD:727812}; Rtn4rl1; rtn4rl1 {ECO:0000312
Gene Symbols Rtn4rl1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NgR3 (aa 361-400).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146760, 237847, 303311
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.