missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTN4RL1 Recombinant Rabbit Monoclonal Antibody (24GB1370)

Description
RTN4RL1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
RTN4RL1 may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.
Specifications
Specifications
| Antigen | RTN4RL1 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 24GB1370 |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Rtn4rl1 |
| Gene Accession No. | Q80WD0, Q86UN2, Q8K0S5 |
| Gene Alias | NgR3; Ngrh2; NGRL2; Nogo receptor-like 2; Nogo-66 receptor homolog 2; Nogo-66 receptor-related protein 3; reticulon 4 receptor like 1; reticulon 4 receptor-like 1; reticulon-4 receptor-like 1; RGD:727812}; Rtn4rl1; rtn4rl1 {ECO:0000312 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?