missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555448
This item is not returnable.
View return policy
Description
Immunogen sequence: YRIHVASRPD SGPHRAEFCE ACQEGIVHWK PSEKLLEEEV TTYTSEASKP RAQAGSGYNF Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.
RTP2 specifically promotes functional cell surface expression of olfactory receptors, but not of other GPCRs.
Specifications
| RTP2 | |
| Polyclonal | |
| Unconjugated | |
| RTP2 | |
| 3CxxC-type zinc finger protein 2; Gm605; receptor (chemosensory) transporter protein 2; receptor transporter protein 2; receptor transporting protein 2; receptor-transporting protein 2; Rtp2; Z3CXXC2; zinc finger, 3CxxC-type 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 344892 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5QGT7 | |
| RTP2 | |
| Recombinant protein corresponding to Human RTP2. Recombinant protein control fragment (Product #RP-90583). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction