missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RTP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562263
This item is not returnable.
View return policy
Description
Immunogen sequence: PSSLTSIFTN TLSEPTDGPV ATKEASITFP FIFTDVKDAV AEVAEGNGKE GGGQGLVPVG HDALPETNAG GLPSQVKGSL ALPFPADVQG KDAFTD Highest antigen sequence identity to the following orthologs: Mouse - 23%, Rat - 26%.
RTP5 gene ontology annotations related to this gene include detection of chemical stimulus involved in sensory perception of bitter taste; protein insertion into membrane; protein targeting to membrane.
Specifications
| RTP5 | |
| Polyclonal | |
| Unconjugated | |
| RTP5 | |
| 3CxxC-type zinc finger protein 5; C2orf85; CXXC finger protein 11; CXXC11; CXXC-type zinc finger protein 11; receptor (chemosensory) transporter protein 5 (putative); receptor transporter protein 5 (putative); receptor-transporting protein 5; RTP5; Z3CXXC5; zinc finger, 3CxxC-type 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 285093 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14D33 | |
| RTP5 | |
| Recombinant protein corresponding to Human RTP5. Recombinant protein control fragment (Product #RP-99701). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction