Learn More
Abeba™ Rubber 9110 Shoes
Product Code. 15693118
Click to view available options
Size:
36
37
41
45
Unit Size:
1 pair
Product Code. 15693118
Brand: Abeba™ 9110/36
Only null left
Add to basket
Add to basket
Acid and base resistant. Rubber sole provides high slip resistance.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Black
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Leather insole
  • adjustable instep area
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC/li
Color Black
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Rubber
Style 9110