Learn More
Abeba™ Rubber 9300 Shoes
Product Code. 15613238
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15613238
Brand: Abeba™ 9300/45
Only null left
Add to basket
Add to basket
Acid and base resistant. Rubber sole provides high slip resistance.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather, perforated
  • White
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC/li
Color White
Material Leather
Size 45
Sole Features Slip-resistant
Sole Material Rubber
Style 9300