Learn More
Deutsch Neumann™ Rubber Ballon
Click to view available options
Diameter (Metric) Opening:
6 mm
Unit Size:
Each
Specifications
Specifications
| Material | Rubber |
| Pore Size | 6mm |
| Type | Rubber Balloon |
| Includes | With Suction And Discharge Valves |
| Secure Tip Attachment | 5 and 8 |
| Diameter (Metric) Opening | 6 mm |