missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RUFY1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101856
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RUFY1.
This gene encodes a protein that contains a RUN domain and a FYVE-type zinc finger domain. The encoded protein binds to phosphatidylinositol-3-phosphate (PI3P) and plays a role in early endosomal trafficking, tethering and fusion through interactions with small GTPases including Rab4, Rab5 and Rab14. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| RUFY1 | |
| Polyclonal | |
| Unconjugated | |
| RUFY1 | |
| 3000002E04Rik; FYVE-finger containing protein; FYVE-finger protein EIP1; La-binding protein 1; Rab4a interacting protein; rab4-interacting protein; Rabip4; RUFY1; RUN and FYVE domain containing 1; RUN and FYVE domain-containing protein 1; ZFYVE12; zinc finger FYVE domain-containing protein 12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 216724, 80230 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BIJ7, Q96T51 | |
| RUFY1 | |
| A synthesized peptide derived from human RUFY1(Accession Q96T51), corresponding to amino acid residues V459-E509. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction