Learn More
RUFY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19180-1-AP-150UL
 View more versions of this product
Product Code. 16808914
  • € 410.00 / 150µL
Description
RUFY2 (RUN And FYVE Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is RUNDC3B.
Specifications
Specifications
RUFY2
Polyclonal
Unconjugated
RUFY2
KIAA1537, RABIP4R, RUFY2, ZFYVE13
Rabbit
Antigen Affinity Chromatography
RUO
55680, 690777, 70432
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R4C2, Q8WXA3
RUFY2
RUFY2 Fusion Protein Ag5938
150 μL
Primary
Human, Mouse, Rat