missing translation for 'onlineSavingsMsg'
Learn More

RUNDC3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16880075 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16880075 150 μL 150µL
16890075 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16880075 Supplier Proteintech Supplier No. 205311AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

May act as an effector of RAP2A in neuronal cells.

Specifications

Antigen RUNDC3A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RUNDC3A
Gene Accession No. O08576, Q59EK9
Gene Alias Rap2 interacting protein 8, RAP2IP, RPIP 8, RPIP8, RUN domain containing 3A, RUNDC3A
Gene Symbols Rundc3a
Host Species Rabbit
Immunogen RUNDC3A Fusion Protein Ag14393
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10900, 303569, 51799
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.