missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX1, Mouse anti-Human, Clone: 1B4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00000861-M19.100ug
This item is not returnable.
View return policy
Description
Sequence: RYHTYLPPPYPGSSQAQGGPFQASSPSYHLYYGASAGSYQFSMVGGERSPPRILPPCTNASTGSALLNPSLPNQSDVVEAEGSHSNSPTNMAPSARLEEAVWRPYSpecifications
| RUNX1 | |
| Monoclonal | |
| Unconjugated | |
| runt-related transcription factor 1 | |
| AML1/AML1-EVI-1/AMLCR1/CBFA2/EVI-1/PEBP2aB | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1B4 | |
| In 1x PBS, pH 7.4 | |
| NM_001754 | |
| RUNX1 | |
| RUNX1 (NP_001745, 376 a.a. ∼ 480 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 861 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction