missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX1 (N-terminal) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19555-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody (19555-1-AP) does not recognize 37 kDa isoform 11.
RUNX1 belongs to the mammalian RUNX family of DNA binding proteins that regulate the expression of genes involved in cellular differentiation and cell cycle progression. RUNX1 is essential for hematopoietic stem cell generation in the vascular regions of the aorta, vitelline and umbilical arteries, yolk sac and placenta. RUNX1 is expressed in all tissues except brain and heart, with highest levels in thymus, bone marrow and peripheral blood.Specifications
| RUNX1 (N-terminal) | |
| Polyclonal | |
| Unconjugated | |
| RUNX1 | |
| AML1, AML1 EVI 1, AMLCR1, CBF alpha 2, CBFA2, EVI 1, Oncogene AML 1, PEA2 alpha B, PEBP2 alpha B, PEBP2aB, RUNX1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12394, 50662, 861 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q01196, Q03347, Q63046 | |
| Runx1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction