missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RUNX1/RUNX1T1 Polyclonal Antibody

Description
RUNX1/RUNX1T1 Polyclonal Antibody for Western Blot, ChIP, ChIP-seq, ELISA
AML1/Runx1 binds DNA as a monomer and through the Runt domain. DNA binding is increased by heterodimerization with CBFB. Isoform AML1L can neither bind DNA nor heterodimerize and interferes with the transactivation activity of AML1/Runx1. CBF binds to the core site, 5'-PYGPYGGT-3', of a number of enhancers and promoters, including murine leukemia virus, polyomavirus enhancer, T cell receptor enhancers, LCK, IL3 and GMCSF promoters. The alpha subunit binds DNA and appears to have a role in the development of normal hematopoiesis. AML1/Runx1 is expressed in all tissues examined except brain and heart, and is expressed at the highest levels in thymus, bone marrow and peripheral blood. Defects in AML1/Runx1 are the cause of familial platelet disorder with associated myeloid malignancy, an autosomal dominant disease characterized by qualitative and quantitative platelet defects, and propensity to develop acute myelogenous leukemia.
Specifications
Specifications
| Antigen | RUNX1/RUNX1T1 |
| Applications | ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 0.05% sodium azide |
| Gene | RUNX1 |
| Gene Accession No. | Q01196, Q06455 |
| Gene Alias | acute myelogenous leukemia 1 translocation 1, cyclin-D related; acute myeloid leukemia 1 protein; AML1; aml1 oncogene; AML1-EVI-1; AML1-EVI-1 fusion protein; AML1-MTG8; AML1T1; AMLCR1; CBF2alpha; CBFA2; CBFA2T1; CBFA2T1 isoform r1t1-11a61; CBFA2T1 isoform r1t1-11a62; CBFA2T1 isoform r1t1-11a63; CBFA2T1 isoform r1t1-11a64; CBFA2T1 isoform r1t1-11a65; CBFA2T1 isoform r1t1-7a47; CBFA2T1 isoform r1t1-7a48; CBFA2T1 isoform r1t1-7a49; CBFA2T1 isoform r1t1-7a50; CBFA2T1 isoform r1t1-7a51; CBFA2T1 isoform r1t1-7a52; CBFA2T1 isoform r1t1-7d53; CBFA2T1 isoform r1t1-7d54; CBFA2T1 isoform r1t1-7d55; CBFA2T1 isoform r1t1-7d56; CBFA2T1 isoform r1t1-8a57; CBFA2T1 isoform r1t1-8a58; CBFA2T1 isoform r1t1-8a59; CBFA2T1 isoform r1t1-8a60; CBF-alpha-2; CDR; Core-binding factor subunit alpha-2; core-binding factor, runt domain, alpha subunit 2; core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclin D-related; Cyclin-D-related protein; Eight twenty one protein; ETO; EVI-1; MTG8; myeloid translocation gene on 8q22; oncogene AML-1; PEA2-alpha B; PEBP2aB; PEBP2alpha; PEBP2-alpha B; polyomavirus enhancer-binding protein 2 alpha B subunit; Protein CBFA2T1; Protein ETO; Protein MTG8; runt related transcription factor 1; runt related transcription factor 1; translocated to, 1 (cyclin D related); runt-related transcription factor 1; runt-related transcription factor 1; translocated to, 1 (cyclin D-related); RUNX1; RUNX1 translocation partner 1; RUNX1/ZNF687 fusion; RUNX1T1; SL3/AKV core-binding factor alpha B subunit; SL3-3 enhancer factor 1 alpha B subunit; transcription factor; zinc finger MYND domain-containing protein 2; ZMYND2 |
| Gene Symbols | RUNX1, RUNX1T1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?