missing translation for 'onlineSavingsMsg'
Learn More

RUNX2/CBFA1 Antibody (3F5), Novus Biologicals™

Product Code. 18388447 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388447 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18388447 Supplier Novus Biologicals Supplier No. H00000860M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RUNX2/CBFA1 Monoclonal antibody specifically detects RUNX2/CBFA1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Functional Assay, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX2/CBFA1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Functional Assay, ChIP Assay
Classification Monoclonal
Clone 3F5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Functional, Chromatin Immunoprecipitation (ChIP)
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004339
Gene Alias Acute myeloid leukemia 3 protein, CBFA1, CBF-alpha-1, CCD1, CCDAML3, CLCD, Core-binding factor subunit alpha-1, core-binding factor, runt domain, alpha subunit 1, MGC120023, ML3, oncogene AML-3, OSF2, OSF-2, osteoblast-specific transcription factor 2, PEA2aA, PEA2-alpha A, PEBP2A, PEBP2aA, PEBP2-alpha A, polyomavirus enhancer-binding protein 2 alpha A subunit, runt domain, alpha subunit 1, runt related transcription factor 2, runt-related transcription factor 2, RUNX2, SL3/AKV core-binding factor alpha A subunit, SL3-3 enhancer factor 1 alpha A subunit
Host Species Mouse
Immunogen RUNX2 (NP_004339, 251 a.a. ~ 350 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NPRPSLNSAPSPFNPQGQSQITDPRQAQSSPPWSYDQSYPSYLSQMTSPSIHSTTPLSSTRGTGLPAITDVPRRISDDDTATSDFCLWPSTLSKKSQAGA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Epigenetics, Growth and Development, Stem Cells, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 860
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.