missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX3 Monoclonal Antibody (OTI2G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810575
This item is not returnable.
View return policy
Description
This gene encodes a member of the runt domain-containing family of transcription factors. A heterodimer of this protein and a beta subunit forms a complex that binds to the core DNA sequence 5′-PYGPYGGT-3′ found in a number of enhancers and promoters, and can either activate or suppress transcription. It also interacts with other transcription factors. It functions as a tumor suppressor, and the gene is frequently deleted or transcriptionally silenced in cancer. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| RUNX3 | |
| Monoclonal | |
| Unconjugated | |
| RUNX3 | |
| AML2, CBFA3, PEBP2aC | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 864 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13761 | |
| RUNX3 | |
| Full length human recombit protein of human RUNX3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction