missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUSC2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81786MFV500
This item is not returnable.
View return policy
Description
RUSC2 Polyclonal antibody specifically detects RUSC2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RUSC2 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| interacting protein of Rab1, Iporin, KIAA0375, RUN and SH3 domain containing 2, RUN and SH3 domain-containing protein 2 | |
| RUSC2 antibody was raised against an 18 amino acid synthetic peptide from near the amino terminus of human RUSC2. The immunogen is located within amino acids 70 - 120 of RUSC2. Amino Acid Squence: TSKRSRDGRGPGAPKRHN | |
| 0.1 mL | |
| Signal Transduction | |
| 9853 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction