Learn More
RuvA Antibody (RuvA 12C6), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50278AF532
This item is not returnable.
View return policy
Description
RuvA Monoclonal antibody specifically detects RuvA in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| RuvA | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A purified | |
| Epigenetics | |
| E. coli | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry | |
| RuvA 12C6 | |
| Western Blot, ELISA, Immunohistochemistry | |
| EC 3.1.22.4, Holliday junction ATP dependent DNA helicase ruvA, Ruv A | |
| Ruv A protein from E. Coli | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |