missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RuvA Antibody (RuvA 12C6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50278CL1
This item is not returnable.
View return policy
Description
RuvA Monoclonal antibody specifically detects RuvA in E. coli samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Radioimmune Assays
Specifications
| RuvA | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 3.1.22.4, Holliday junction ATP dependent DNA helicase ruvA, Ruv A | |
| Ruv A protein from E. Coli | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Radioimmune Assays (RIA) | |
| RuvA 12C6 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| E. coli | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction