missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RUVBL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17928942
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17928942 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17928942 Supplier Invitrogen™ Supplier No. PA5144643

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive control: Hela cell lysate, MCF-7 cell lysate, NIH/3T3 cell lysate, mouse liver tissue lysate, mouse skeletal muscle tissue lysate, rat skin tissue lysate, Rat skeletal muscle lysate, human liver tissue, human testis tissue, rat trachea tissue, human fallopian tube tissue, mouse skeletal muscle tissue. Predicted band size: 50 kDa Subcellular Location: Cytoplasm, Cytoskeleton, Membrane, Nucleus.

The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUVBL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene RUVBL1
Gene Accession No. P60122, P60123, Q9Y265
Gene Alias 2510009G06Rik; 49 kDa TATA box-binding protein-interacting protein; 49 kDa TBP-interacting protein; 54 kDa erythrocyte cytosolic protein; DNA helicase p50; ECP54; ECP-54; INO80 complex subunit H; INO80H; NMP 238; NMP238; Nuclear matrix protein 238; PONTIN; Pontin 52; Pontin52; RuvB (E coli homolog)-like 1; RuvB like AAA ATPase 1; Ruvbl1; ruvB-like 1; RuvB-like AAA ATPase 1; RuvB-like protein 1; RVB1; TAP54-alpha; TATA binding protein interacting protein 49 kDa; TIH1; Tip49; TIP49A; TIP60-a; TIP60-associated protein 54-alpha
Gene Symbols RUVBL1
Host Species Rabbit
Immunogen Recombinant protein within Human TIP49A aa 121-320/456.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56505, 65137, 8607
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.