missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RuvC Antibody (RuvC 5G9/3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50280AF405
This item is not returnable.
View return policy
Description
RuvC Monoclonal antibody specifically detects RuvC in E. coli samples. It is validated for Western Blot, ELISA.
Specifications
| RuvC | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Ruv C protein from E. Coli. | |
| 0.1 mL | |
| DNA Repair, Epigenetics | |
| E. coli | |
| Purified |
| Western Blot, ELISA | |
| RuvC 5G9/3 | |
| Western Blot, ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction