Specifications
| Type | Distilling Spider |
| For Use With (Application) | For simultaneous distillation in 20 Distilling sleeves |
| Quantity | 1 Pack |
| Standard Taper Joint Side | NS 29/32 |
| Volume (Metric) | 20 mL |