missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RWDD3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76309MFV450
This item is not returnable.
View return policy
Description
RWDD3 Polyclonal antibody specifically detects RWDD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| RWDD3 | |
| Polyclonal | |
| Western Blot, ELISA | |
| DKFZp566K023, FLJ51225, RSUME, RWD domain containing 3, RWD domain-containing protein 3, RWD-containing sumoylation enhancer | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human RWDD3. The immunogen is located within the first 50 amino acids of RWDD3. Amino Acid Squence: CRPHEWEVLSRSETDGT | |
| 0.1 mL | |
| Innate Immunity | |
| 25950 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction