missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RWDD3 Monoclonal Antibody (OTI2A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811762S
This item is not returnable.
View return policy
Description
RWDD3 Monoclonal antibody specifically detects RWDD3 in Human samples. It is validated for Western BlotSpecifications
| RWDD3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11310 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 34 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2A4 | |
| Unconjugated | |
| ACADM | |
| 2510027J23Rik; 3110037C01Rik; AI035684; AI662458; pituitary tumor X2CR1 protein; RSUME; RWD domain containing 3; RWD domain-containing protein 3; RWD domain-containing sumoylation enhancer; RWD-containing sumoylation enhancer; Rwdd3; X2cr1 | |
| Full length human recombinant protein of human ACADM produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction