missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RWDD4A Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97777AF700
This item is not returnable.
View return policy
Description
RWDD4A Polyclonal specifically detects RWDD4A in Human samples. It is validated for Western Blot.
Specifications
| RWDD4A | |
| Polyclonal | |
| Western Blot | |
| FAM28A, member A, MGC10198, Protein FAM28A, RWD domain containing 4, RWD domain containing 4A, RWD domain-containing protein 4, RWD domain-containing protein 4A, RWDD4A | |
| Produced in rabbits immunized with E. coli-derived Human RWDD4A fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 201965 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction