missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXR gamma/NR2B3 Antibody (PCRP-RXRG-5H4) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14170JF635
This item is not returnable.
View return policy
Description
RXR gamma/NR2B3 Monoclonal antibody specifically detects RXR gamma/NR2B3 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| RXR gamma/NR2B3 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| NR2B3retinoid X receptor-gamma, Nuclear receptor subfamily 2 group B member 3, retinoic acid receptor RXR-gamma, Retinoid X receptor gamma, retinoid X receptor, gamma, RXRC | |
| Recombinant full-length human RXR gamma/NR2B3 protein (Uniprot: P48443) | |
| 0.1 mL | |
| Cancer, Transcription Factors and Regulators | |
| 6258 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunoprecipitation, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-RXRG-5H4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction