missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RXRA Recombinant Rabbit Monoclonal Antibody (23GB2815), Invitrogen™
GREENER_CHOICE

Product Code. 30089889
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30089889 100 μL 100µL
30087635 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30089889 Supplier Invitrogen™ Supplier No. MA553566

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXRA
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2815
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rxra
Gene Accession No. P19793, P28700, Q05343
Gene Alias 9530071D11Rik; Nr2b1; Nuclear receptor subfamily 2 group B member 1; retinoic acid receptor RXR-alpha; retinoid X nuclear receptor alpha; retinoid X receptor; retinoid X receptor alpha; retinoid X receptor, alpha; rxr alpha; RXR alpha 1; Rxra; RXRalpha1
Gene Symbols Rxra
Host Species Rabbit
Immunogen A synthesized peptide derived from human Retinoid X Receptor alpha (1-41AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20181, 25271, 6256
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.