missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXRG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11129-1-AP-150UL
This item is not returnable.
View return policy
Description
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells (Joshi et al., 2005). The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer (Abu et al., 2005). RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors (Lane and Bailey, 2005).Specifications
| RXRG | |
| Polyclonal | |
| Unconjugated | |
| RXRG | |
| NR2B3, Retinoid X receptor gamma, retinoid X receptor, gamma, RXRC, RXRG | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20183, 6258, 83574 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P28705, P48443, Q5BJR8 | |
| RXRG | |
| RXRG Fusion Protein Ag1611 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction