missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ryanodine Receptor 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95111-0.1ml
This item is not returnable.
View return policy
Description
Ryanodine Receptor 2 Polyclonal antibody specifically detects Ryanodine Receptor 2 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Ryanodine Receptor 2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| arrhythmogenic right ventricular dysplasia 2, ARVC2, ARVD2, cardiac muscle ryanodine receptor-calcium release channel, Cardiac muscle-type ryanodine receptor, cardiac-type ryanodine receptor, hRYR-2, islet-type ryanodine receptor, kidney-type ryanodine receptor, ryanodine receptor 2, ryanodine receptor 2 (cardiac), RyR2, RYR-2, type 2 ryanodine receptor, VTSIP | |
| A synthetic peptide corresponding to a sequence within amino acids 4868-4967 of human Ryanodine Receptor 2 (NP_001026.2). EHTGQESYVWKMYQERC | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Neuroscience, Signal Transduction | |
| 6262 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction