Learn More
Ryanodine Receptor Antibody (34C), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB300-543APC
Product Code. 18505934
  • ONLINE EXCLUSIVE PRICE
    € 582.00
    € 550.20 / 0.10mL
    Save € 31.80
    5% Off
Description
Ryanodine Receptor Monoclonal antibody specifically detects Ryanodine Receptor in Human, Mouse, Rat, Amphibian, Bovine, Canine, Fish, Ferret, Mink, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
Mouse
6261
APC
Partially purified chicken pectoral muscle ryanodine receptor.
Mouse, Amphibian