missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RYBP/DEDAF Antibody (OTI2B4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73962CL1
This item is not returnable.
View return policy
Description
RYBP/DEDAF Monoclonal antibody specifically detects RYBP/DEDAF in Canine,Human,Monkey,Mouse samples. It is validated for Western Blot
Specifications
| RYBP/DEDAF | |
| Monoclonal | |
| CoraFluor 1 | |
| AAP1, APAP-1, Apoptin-associating protein 1, Death effector domain-associated factor, DEDAFYY1 and E4TF1-associated factor 1, RING1 and YY1 binding protein, RING1 and YY1-binding protein, ring1 interactor RYBP, YEAF1DED-associated factor, YY1 and E4TF1 associated factor 1 | |
| Full length human recombinant protein of human RYBP(NP_036366) produced in E.coli. | |
| 0.1 mL | |
| Apoptosis | |
| 23429 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot | |
| OTI2B4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction