missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RYBP/DEDAF Antibody (OTI2B4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73962FR
This item is not returnable.
View return policy
Description
RYBP/DEDAF Monoclonal specifically detects RYBP/DEDAF in Human, Mouse, Canine, Monkey samples. It is validated for Western Blot.
Specifications
| RYBP/DEDAF | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| RYBP | |
| Full length recombinant protein of human RYBP/DEDAF (NP_036366) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Canine, Monkey | |
| IgG1 |
| Western Blot | |
| OTI2B4 | |
| Western Blot | |
| AAP1, APAP-1, Apoptin-associating protein 1, Death effector domain-associated factor, DEDAFYY1 and E4TF1-associated factor 1, RING1 and YY1 binding protein, RING1 and YY1-binding protein, ring1 interactor RYBP, YEAF1DED-associated factor, YY1 and E4TF1 associated factor 1 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis | |
| 23429.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction