missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RYBP Monoclonal Antibody (OTI2H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800035S
This item is not returnable.
View return policy
Description
RYBP Monoclonal antibody specifically detects RYBP in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| RYBP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N488 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23429 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI2H9 | |
| Unconjugated | |
| Rybp | |
| 2410018J24Rik; AAP1; APAP-1; apoptin-associating protein 1; Death effector domain-associated factor; DEDAF; DED-associated factor; RING1 and YY1 binding protein; RING1 and YY1-binding protein; ring1 interactor RYBP; Rybp; YEAF1; YY1 and E4TF1 associated factor 1; YY1 and E4TF1-associated factor 1 | |
| Full length human recombinant protein of human RYBP produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction